Aluminium oxide, α-phase ≥99.997% (metals basis)

Supplier: Thermo Scientific Chemicals

047261-14 047261-22 047261-36
AA47261-14EA 112.97 CAD
AA47261-14 AA47261-22 AA47261-36
Aluminium oxide, α-phase ≥99.997% (metals basis)
Aluminium oxide, α-phase

CAS No.: 1344-28-1 Molecular Formula: Al2O3 Formula Weight: 101.96 Physical Form: 40 Micron Crystalline Density: 3.965 Appearance: White Odor: Odorless Melting Point: 2045° Boiling Point: 2980° Merck Reference: 14,356 MDL No.: MFCD00003424

Formula: Al₂O₃
Boiling Pt: 2980 °C
Melting Pt: 2045 °C
Density: 3.965
Storage Temperature: Ambient
MDL Number: MFCD00003424
CAS Number: 1344-28-1
EINECS: 215-691-6
Merck Index: 14,356

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR