Diaion® PA308 (Cl) 1.0 meq/ml on poly(styrene-divinylbenzene), 4% cross-linked, strongly basic porous type ion-exchange resin I
Supplier: Thermo Scientific Chemicals
046990-A1
AA46990-A1EA 153.1 CAD
AA46990-A1
Diaion® PA308 (Cl) 1.0 meq/ml on poly(styrene-divinylbenzene), 4% cross-linked, strongly basic porous type ion-exchange resin I
Diaion® PA308 (Cl)
Physical Form: 300-1180ηm Beads Appearance: White to yellow
Sugar decolorization