Diaion® HP20, highly porous type synthetic adsorbent resin
Supplier: Thermo Scientific Chemicals
046488-A1
AA46488-A1EA 426.08 CAD
AA46488-A1
Diaion® HP20, highly porous type synthetic adsorbent resin
Diaion® HP20
Physical Form: ≥250ηm Beads Density: 1.01
Amino acid separations, small molecules in medium hydrophobicity in polar solvents