Cerium, sputtering target, Ø 76.2 mm, Thickness: 3,18 mm

Supplier: Thermo Scientific Chemicals
Danger

045926-KS
AA45926-KSEA 2126.68 CAD
AA45926-KS
Cerium, sputtering target, Ø 76.2 mm, Thickness: 3,18 mm
Cerium

CAS No.: 7440-45-1 Odor: Odorless UN No.: 1333 MDL No.: MFCD00010924

Formula: Ce
MW: 140.12 g/mol
Boiling Pt: 3468
Melting Pt: 795
Storage Temperature: Ambient
MDL Number: MFCD00010924
CAS Number: 7440-45-1
EINECS: 231-154-9
UN: 1333
ADR: 4.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR