Sulfur in Isooctane Standard Solutions, Specpure®, Thermo Scientific

Supplier: Thermo Scientific Chemicals

045352-AE
AA45352-AEEA 177.12 CAD
AA45352-AE
Sulfur in Isooctane Standard Solutions, Specpure®, Thermo Scientific
Standards Petroleum Industry Standards

Physical Form: Liquid
Appearance: Colorless
Odor: Petrol-like
UN No.: 1262
MDL No.: MFCD00804269

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR