Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3-4mm (0.12-0.16in)

Supplier: Thermo Scientific Chemicals

Synonyms: Linde 13X

033550-A1 033550-A7
AA33550-A1EA 142.39 CAD
AA33550-A1 AA33550-A7
Molecular sieve 13X (1.0 nm, 10 Å), pellets, Ø 3-4mm (0.12-0.16in)
Molecular sieve 13X (1.0 nm, 10 Å)

1/8in Pellets


CAS No.: 63231-69-6 Physical Form: 1/8in Pellets Sensitivity: Hygroscopic MDL No.: MFCD00131613

Storage Temperature: Ambient
MDL Number: MFCD00164506
CAS Number: 63231-69-6

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR