Avantor® Seradigm, Ultimate Grade Fetal Bovine Serum (FBS)
Supplier: VWR
1600-500G 1600-500GH 1600-050 1600-050H 1600-500 1600-500H
97068-102EA 1808.75 CAD
97068-102 97068-104 89510-198 89510-200 97068-101 97068-107
Avantor® Seradigm, Ultimate Grade Fetal Bovine Serum (FBS)
Sera (Serum)
Ultimate Grade FBS is the ultimate in product quality and viral safety, undergoing additional BVD virus testing using qPCR technology.
  • 100% US Origin
  • Additional virus testing through qPCR technology
  • Enhanced traceability and regulatory compliance
  • ISIA Certified Supply Chain for Traceability
Industry-leading collection and production techniques provide additional features that elevate product quality, enhance traceability, and improve regulatory compliance.
Avantor Seradigm is a supplier of animal sera that provides the cell culture community withMore Product Information