VWR® Vacuum Filtration Systems featuring Pall Membranes
Supplier: VWR International
97066-200 97066-202 97066-206 97066-208 97066-212 97066-214 97066-216 97066-218 97066-220 97066-222 97066-232 97066-234 97066-204 97066-210 89220-694 89220-696 89220-698 89220-700 89220-702 89220-704 89220-708 89220-714 89220-716 89220-724 89220-726 89220-728
97066-200CS 396.77 CAD
97066-200 97066-202 97066-206 97066-208 97066-212 97066-214 97066-216 97066-218 97066-220 97066-222 97066-232 97066-234 97066-204 97066-210 89220-694 89220-696 89220-698 89220-700 89220-702 89220-704 89220-708 89220-714 89220-716 89220-724 89220-726 89220-728
VWR® Vacuum Filtration Systems featuring Pall Membranes
Filters Bottle Top Filters
VWR® Vacuum Filtration Systems are designed for the preparation of buffers, tissue culture media, microbiological media, and other biological fluids.
  • For the preparation of biological fluids
  • Feature Pall membranes
  • Compatible with the quick-connect vacuum pedestal for use with existing laboratory stands
They feature large diameter Pall membranes for guaranteed performance and maximum throughput.
The centrifuge tube filtration system includes a filter funnel, 15 or 50 ml centrifuge tube with polypropylene screw cap, and vacuum hose adapter. TheMore Product Information