nUView™ Tris-Glycine Precast Gels, NuSep®
Supplier: NuSep
NG21-008 NG21-010 NG21-012 NG21-420 NG21-816 NG11-010 NG11-012 NG11-420 NG31-420 NB10-008 NB10-010 NB10-012 NB10-816 NB12-008 NB12-010 NB12-012 NB12-420 NB12-816 NB17-008 NB17-010 NB17-012 NB17-420 NB17-816 NN10-008 NN10-010 NN10-012 NN10-420 NN10-816 NN12-008 NN12-010 NN12-012 NN12-420 NN12-816 NN17-008 NN17-010 NN17-012 NN17-420 NN17-816 NG31-010 NG31-012 NG31-010 NG31-012 NG11-008 NG11-816
81002-000PK 358.66 CAD
81002-000 81002-002 81002-004 81002-006 81002-008 81002-070 81002-072 81002-074 81002-084 97065-116 97065-118 97065-120 97065-122 97065-124 97065-126 97065-128 97065-130 97065-132 97065-134 97065-136 97065-138 97065-140 97065-142 97065-144 97065-146 97065-148 97065-150 97065-152 97065-154 97065-156 97065-158 97065-160 97065-162 97065-164 97065-166 97065-168 97065-170 97065-172 97065-174 470128-556 470128-558 490012-290 490012-292 490015-608 490015-610
nUView™ Tris-Glycine Precast Gels, NuSep®
Electrophoresis Gels Precast Gels
Precast gels incorporate a unique formulation allowing protein bands to be visualized in only two minutes in the presence of ultraviolet (UV) light
The nUView technology increases protein recovery and saves significant time, since gels do not require staining and de-staining. Cassettes have large sample well capacities (up to 50μL). Numbered lanes and solid well dividers offer easy well identification to eliminate cross sample contamination and gel damage. Separations can be carried out under native or denaturing conditions using a broad range of buffers including MES and MOPS. EachMore Product Information