VWR® TBS Buffer, 20X Liquid
Supplier: VWR
J640-4L
97064-338EA 480.65 CAD
97064-338
VWR® TBS Buffer, 20X Liquid
Buffers Electrophoresis Buffers Washing Buffers
A 1X solution of Tris Buffered Saline (TBS) contains 140 mM Sodium Chloride, 3.0 mM Potassium Chloride and 25 mM Tris.