Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade

Supplier: VWR

Synonyms: Methanesulphinylmethane, Methyl sulfoxide, Dimethyl(oxido)sulfur, Methyl sulphoxide, Dimethyl(oxido)sulphur, DMSO, Methanesulfinylmethane

N182-5X10ML
97063-136EA 290.94 CAD
97063-136
Dimethyl sulfoxide ≥99.9%, Ultra Pure Grade
Dimethyl sulphoxide
Formula: (CH₃)₂SO
MW: 78.14 g/mol
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18.5 °C
Density: 1.101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=157e2a24c93041cbbb69833d226195e333b07665&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Acidity Test Pass
Congealing Point = 18.3°C
Appearance Clear colorless liquid.
Non-volatile Residue = 5 mg
Product Lock Test C9481
Purity = 99.9 %
Refractive Index 1.4755 - 1.4775
Specific Gravity 1.095 - 1.101
Water (KF) < = 0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR