VWR® MOPS Buffer Solution 10X, Biotechnology Grade
Supplier: VWR
E526-500ML
97062-852EA 142.82 CAD
97062-852 97062-854
VWR® MOPS Buffer Solution 10X, Biotechnology Grade
Buffers Electrophoresis Buffers Running Buffers
Optimized for RNA Agarose Electrophoresis
Ideal for Use with RNA Formaldehyde Gels.
Order Now
SPECIFICATIONS
Environmentally Preferable