Magnesium chloride 1 M aqueous solution, Biotechnology Grade

Supplier: VWR

E525-100ML E525-500ML
97062-848EA 134.32 CAD
97062-848 97062-850
Magnesium chloride 1 M aqueous solution, Biotechnology Grade
Magnesium chloride

A reagent for the preparation of competent cells for transformation.

Formula: MgCl₂
MW: 95.21 g/mol
Boiling Pt: 1412
Melting Pt: 715
Storage Temperature: Ambient
MDL Number: MFCD00011106
CAS Number: 7786-30-3
Merck Index: 13,05684

Order Now

Specification Test Results

Conductivity (1:100) @25C 2000 - 2400 umhos
Identification PASS / Fail
Insolubles ≤ 0.005 %
Nuclease NONE / Fail

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR