Bovine Serum Albumin (BSA)
Supplier: VWR
0903-5G
97062-508EA 320.99 CAD
97062-508
Bovine Serum Albumin (BSA)
Proteins and Peptides
Crystalline. Prepared by a cold alcohol isolation procedure. This salt-free preparation is the purest material ideal for sensitive biochemical and diagnostic assays. DNase- and RNase-free, virtually free of globulins and other interfering contaminants.