Agarose, II™ Biotechnology

Supplier: VWR

0815-250G 0815-25G
97062-396CS 8197.77 CAD
97062-396 97062-398 97062-400
Agarose, II™ Biotechnology
Agarose

A low-melting agarose that melts between 62 to 68 °C and remains liquid for several hours at 37 °C. Ideal for purification of macromolecules from agarose slices and for in-gel enzymatic manipulations.

Storage Temperature: Ambient
MDL Number: MFCD00081294
CAS Number: 9012-36-6
EINECS: 232-731-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=526c20a29610acf6900f898ff245c5602558c785&vid=ae34e4c76a4d7838d4dc9501397f6741b1b97d5e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

DNase NONE
EEO (-Mr) ≤0.12
Gel Strength (1.0%) ≥250 g/cm²
Gelling Range (1.0%) 24 - 29 °C
Melting Range 62 - 68 °C
Moisture < 10 %
Protease NONE
RNase NONE
Sulfate ≤0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR