Lithium dodecyl sulfate ≥99%, high purity

Supplier: VWR
Danger

Synonyms: Dodecyl sulphuric acid lithium salt, Dodecyl sulfate lithium salt, Dodecyl sulphate lithium salt, Lithium lauryl sulphate, Lauryl sulfate lithium salt, Dodecyl sulfuric acid lithium salt, Lithium lauryl sulfate, Lauryl sulphate lithium salt

0782-25G
97062-358EA 248.82 CAD
97062-358
Lithium dodecyl sulfate ≥99%, high purity
Lithium dodecyl sulphate

Anionic - negatively-charged (carboxylates, sulfates or sulfonates in alkali metal form). Utilized for applications ranging from solubilizing membrane proteins in non-denaturing conditions to creating denaturing conditions during gel electrophoresis. Used for complete disruption of membranes.

Formula: C₁₂H₂₅LiO₄S
MW: 272.34 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00007467
CAS Number: 2044-56-6
EINECS: 218-058-2
UN: 1325
ADR: 4.1 III

Order Now

Specification Test Results

Abs.@260nm (5%, Water) = 0.1
Abs.@280nm (5%, Water) = 0.1
Loss on Drying = 3 %
Purity = 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR