EDTA dipotassium salt dihydrate, Ultrapure

Supplier: VWR

Synonyms: Ethylenediaminetetraacetic acid dipotassium salt dihydrate, Titriplex® Potassium salt, Ethylenediaminetetra-acetic acid dipotassium salt dihydrate, Edetic acid dipotassium salt dihydrate, Dipotassium dihydrogen ethylenediaminetetraacetate dihydrate, Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrate, EDTA dipotassium dihydrate, Potassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrate, Ethylenedinitrilotetraacetic acid dipotassium salt

0720-500G
97062-274CS 1342.89 CAD
97062-274
EDTA dipotassium salt dihydrate, Ultrapure
EDTA dipotassium salt dihydrate
Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
Melting Pt: 272
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS Number: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546

Order Now

Specification Test Results

Chloride = 0.005 %
Heavy Metals = 0.001 %
Iron = 0.001 %
pH (5%, Water) @25°C 4.0 - 5.0
Purity = 99.0 %
Appearance White granular powder


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR