Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Ultrapure

Supplier: VWR
Warning

Synonyms: 2,2-Bis(hydroxymethyl)-2,2',2''-nitrilotriethanol, Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS), 2-(Bis(2-hydroxyethyl)amino)-2-(hydroxymethyl)propane-1,3-diol, 2-[Bis(2-hydroxyethyl)amino]-2-(hydroxymethyl)propane-1,3-diol

0715-100G 0715-250G 0715-500G
97062-262CS 1797.65 CAD
97062-262 97062-264 97062-266
Bis(2-hydroxyethyl)amino tris(hydroxymethyl)methane (BIS-TRIS), Ultrapure
Bis(2-hydroxyethyl)amino-tris(hydroxymethyl)methane (BIS-TRIS)

Biological buffers are useful for cell culture in vitro, enzyme assays and some electrophoretic applications at physiological pH. Universally applicable buffers for biochemistry must be water soluble, not interfere with biological processes or biological

Formula: C₈H₁₉NO₅
MW: 209.24 g/mol
Melting Pt: 103…104 °C
Storage Temperature: Ambient
MDL Number: MFCD00002853
CAS Number: 6976-37-0
EINECS: 230-237-7

Order Now

Specification Test Results

DNase none detected
RNase none detected
Melting Point 102 °C - 106 °C
Moisture (KF) = 1 %
pH (1.0%, Water) @25°C 8.8 - 9.6
pKa @25°C 6.45 - 6.65
Protease none
Purity (Titration) 99 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR