Benzamidine hydrochloride hydrate ≥98% (by anhydrous basis), powder, Ultra Pure Grade

Supplier: VWR
Warning

Synonyms: Phenylamidine hydrochloride hydrate, Benzene-1-carboximidamide hydrochloride hydrate, Benzenecarboximidamide hydrochloride hydrate, Benzamidinium chloride hydrate

0616-25G
97062-038EA 252.1 CAD
97062-038
Benzamidine hydrochloride hydrate ≥98% (by anhydrous basis), powder, Ultra Pure Grade
Benzamidine hydrochloride hydrate

A protease inhibitor of arginine-selective enzymes like trypsin, kallikrein and thrombin.


  • Fine white crystalline powder

Formula: C₆H₅C(=NH)NH₂·HCl·H₂O
MW: 174.63 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00066285
CAS Number: 206752-36-5
EINECS: 216-795-4

Order Now

Specification Test Results

Em (Lambda Max, MEOH, anhy) = 9500
Heavy Metals (as Pb) = 0.005 %
Melting Point 77 °C - 87 °C
Purity (Anhydrous) = 98 %
Solubility (1%, Water) pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR