Sodium molybdate(VI) dihydrate ACS

Supplier: VWR

0541-250G
97061-880CS 2857.49 CAD
97061-880
Sodium molybdate(VI) dihydrate ACS
Sodium molybdate(VI) dihydrate
Formula: Na₂MoO₄·2H₂O
Melting Pt: 687 °C
Density: 3.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149170
CAS Number: 10102-40-6
EINECS: 231-551-7
Merck Index: 13,08718

Order Now

Specification Test Results

Ammonium 0.001%
Chloride = 0.005%
Heavy Metals (as Pb) = 0.0005%
Insolubles = 0.005%
Iron = 0.001%
pH (5%, Water) @25°C 7 - 10.5
Phosphate = 0.0005%
Purity 99.5% - 103%
Sulfate = 0.015%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR