Coomassie® brilliant blue R-250, Ultrapure

Supplier: VWR

0472-10G 0472-25G 0472-50G
97061-738EA 121.68 CAD
97061-738 97061-740 97061-742
Coomassie® brilliant blue R-250, Ultrapure
Coomassie® Brilliant Blue R-250

Commonly used stain for the detection of protein bands following electrophoresis.

Formula: C₄₅H₄₄N₃NaO₇S₂
Melting Pt: 174…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00041762
CAS Number: 6104-59-2
EINECS: 228-060-5

Order Now

Specification Test Results

Dye Content (Dry Basis) = 65 %
Em (550nm, Water, Dry Basis) = 33000
Loss on Drying = 5.0 %
Solubility (0.020g/L, Water) Pass


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR