Disodium hydrogen phosphate heptahydrate ACS

Supplier: VWR

0348-12KG 0348-1KG 0348-50KG 0348-2.5KG 0348-500G
97061-466EA 1543.58 CAD
97061-466 97061-468 97061-474 97061-470 97061-472
Disodium hydrogen phosphate heptahydrate ACS
di-Sodium hydrogen phosphate heptahydrate
Formula: HNa₂O₄P·7H₂O
Melting Pt: 48 °C
Density: 1.7 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149180
CAS Number: 7782-85-6
EINECS: 231-448-7

Order Now

Specification Test Results

Chloride 0.001%
Heavy Metals (as Pb) = 0.001%
Insolubles = 0.005%
Iron = 0.001%
pH (5%, Water) @25°C 8.7 - 9.3
Purity (as is) 98% - 102%
Sulfate = 0.005%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR