Spectra/Por® Float-A-Lyzer G2 Dialysis Devices, Spectrum® Laboratories
Supplier: Spectrum Laboratories
G235033 G235057 G235025 G235049 G235061 G235027 G235051 G235063 G235029 G235053 G235065 G235031 G235055 G235067 G235069 G235034 G235058 G235070 G235035 G235059 G235071 G235036 G235060 G235072 G235037 G235062 G235073
28170-336PK 432.34 CAD
28170-336 28170-362 97059-000 97059-002 97059-004 97059-006 97059-008 97059-010 97059-012 97059-014 97059-016 97059-018 97059-020 97059-022 97059-024 97059-026 97059-028 97059-030 97059-032 97059-034 97059-036 97059-038 97059-040 97059-042 97059-044 97059-046 97059-048
Spectra/Por® Float-A-Lyzer G2 Dialysis Devices, Spectrum® Laboratories
Tubing Dialysis Tubing
Spectrum introduces the Spectra/Por® Float-A-Lyzer® G2, the next generation in Ready-to-Use laboratory dialysis devices featuring proprietary Ultra-pure Biotech Cellulose Ester (CE) Membrane.
Spectrum's proprietary Biotech Grade CE is a low protein-binding synthetic membrane available in 9 precise MWCO's with no heavy metal and sulfide contaminants. The cylindrical tubing geometry prevents sample dilution (associated with cassette-type devices) and provides open access for total volume retrieval by pipette. Only the Float-A-Lyzer G2 assures a 95 to 98% sample recovery while maintaining 99%More Product Information