Stainless Steel Flat Top Worktable with Undershelf, 14-Gauge Type 304, Eagle MHC™
Supplier: Eagle Group
T3030SE T2448SE T2460SE T2472SE T3060SE T3072SE T2424SE T2430SE T2436SE T2484SE T3024SE T3036SE T3048SE T3084SE T3636SE T3648SE T3660SE T3672SE T3684SE
58202-426EA 4252.08 CAD
58202-426 58202-480 58202-488 58202-496 58202-504 58202-512 97058-700 97058-702 97058-704 97058-706 97058-708 97058-710 97058-712 97058-714 97058-716 97058-718 97058-720 97058-722 97058-724
Stainless Steel Flat Top Worktable with Undershelf, 14-Gauge Type 304, Eagle MHC™
Furniture Tables and Workbenches
Tables have a 14 gauge type 304 stainless steel flat top.
  • Non-adjustable
  • Shipped knockdown
  • For use with Eagle MHC worktables only
  • Overshelves are available in single or double style
  • Mounted on four stainless steel 1" Ø tubular base legs
1⁵/₈" Ø legs and adjustable undershelf are heavy gauge stainless steel. Rolled edge front and rear with ends turned under 90 degrees. 35¹/₈ to 36¹/₈" adjustable working height.
Ordering information: Available accessories include drawers and casters. It is recommended that overshelves be ordered when ordering tables.More Product Information