Sample Loops for Rheodyne Sample Injectors, Microsolv Tech
Supplier: MicroSolv Technology Corp
47003-100 47001-100 47003-010 47001-010 47001-001 47003-020 47001-020 47001-002 47003-050 47001-050 47003-005 47001-005
97058-052EA 97.58 CAD
97058-052 97058-056 97058-060 97058-064 97058-068 97058-072 97058-076 97058-080 97058-084 97058-088 97058-092 97058-096
Sample Loops for Rheodyne Sample Injectors, Microsolv Tech
Chromatography Supplies Chromatography Sample Loops
These HPLC injector sample loops are available with 1/16" O.D. Ultra-T™ PEEK tubing or with welded and annealed 316 stainless steel tubing. These options provide unsurpassed injection reproducibility and accuracy.
  • A label with the loop volume is attached
  • Use these loops with Rheodyne ® or Cheminert® brand HPLC injector valves
PEEK versions are supplied with (2) high pressure, PEEK, three-piece, small hex head nuts with 1/32" threads and (2) special, single cone ferrules per nut to avoid twisting and turning of the sample loop. At the same time, theMore Product Information