EXP®2 Stem Trap Kits and Replacement Stems, 3.0 µm EXP
Supplier: Optimize
15-03992-HG 15-03992-HH 15-03992-HI 15-03992-HJ 15-03992-HK 15-03992-HL 15-03992-HM 15-04001-HG 15-04001-HH 15-04001-HI 15-04001-HJ 15-04001-HK 15-04001-HL 15-04001-HM 15-04008-HG 15-04008-HH 15-04008-HI 15-04008-HJ 15-04008-HK 15-04008-HL 15-04008-HM 15-04014-HG 15-04014-HH 15-04014-HI 15-04014-HJ 15-04014-HK 15-04014-HL 15-04014-HM 15-04020-HG 15-04020-HH 15-04020-HI 15-04020-HJ 15-04020-HK 15-04020-HL 15-04020-HM 15-03997-HG 15-03997-HH 15-03997-HI 15-03997-HJ 15-03997-HK 15-03997-HL 15-03997-HM 15-04003-HG 15-04003-HH 15-04003-HI 15-04003-HJ 15-04003-HK 15-04003-HL 15-04003-HM 15-04009-HG 15-04009-HH 15-04009-HI 15-04009-HJ 15-04009-HK 15-04009-HL 15-04009-HM 15-04015-HG 15-04015-HH 15-04015-HI 15-04015-HJ 15-04015-HK 15-04015-HL 15-04015-HM 15-04021-HG 15-04021-HH 15-04021-HI 15-04021-HJ 15-04021-HK 15-04021-HL 15-04021-HM
97056-080PK 2043.64 CAD
97056-080 97056-082 97056-084 97056-086 97056-088 97056-090 97056-092 97056-094 97056-096 97056-098 97056-100 97056-102 97056-104 97056-106 97056-108 97056-110 97056-112 97056-114 97056-116 97056-118 97056-120 97056-122 97056-124 97056-126 97056-128 97056-130 97056-132 97056-134 97056-136 97056-138 97056-140 97056-142 97056-144 97056-146 97056-148 97056-346 97056-348 97056-350 97056-352 97056-354 97056-356 97056-358 97056-360 97056-362 97056-364 97056-366 97056-368 97056-370 97056-372 97056-374 97056-376 97056-378 97056-380 97056-382 97056-384 97056-388 97056-390 97056-392 97056-394 97056-396 97056-398 97056-400 97056-402 97056-406 97056-408 97056-410 97056-412 97056-414 97056-416 97056-418
EXP®2 Stem Trap Kits and Replacement Stems, 3.0 µm EXP
Chromatography Columns
The entire EXP®2 stem trap and reusable holder are only slightly larger than a standard HPLC fitting. Its slim architecture allows it to easily fit into crowded instrument compartments or to connect directly to tightly-spaced injection ports.
  • Rated to 20000+ psi (1400+ bar)
  • Hand-tight and wrench-tight configurationwith auto-adjusting ZDV connection
  • Low-volume/low-dispersion cartridges
  • For nano flow applications
When tightened by hand, the EXP2 stem trap seals to 8700+ psi. All configurations incorporate wrench-flats to enable flawless sealing to 20000+More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable