Rack/Filling Station, for Headspace Vials, Acrylic, MicroSolv™
Supplier: MicroSolv Technology Corp
95025-R
97055-006EA 303.68 CAD
97055-006
Rack/Filling Station, for Headspace Vials, Acrylic, MicroSolv™
Racks Chromatography Vial Racks
The vial racks/filling stations are made of acrylic material so you can see into the vial as you fill it
Also, the silicone base makes them an excellent filling station as they will not slide on the bench.