WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and Snap Caps, Unassembled, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
07-1220B-101 07-1220G-101 07-1220N-101 07-1220R-101 07-1221B-101 07-1221G-101 07-1221N-101 07-1221R-101 07-1245B-101 07-1245G-101 07-1245N-101 07-1245R-101 07-1275B-101 07-1275G-101 07-1275N-101 07-1275R-101 07-2020B-201 07-2020G-201 07-2020N-201 07-2020R-201 07-2021B-201 07-2021G-201 07-2021N-201 07-2021R-201 07-2045B-201 07-2045G-201 07-2045N-201 07-2045R-201 07-2075B-201 07-2075G-201 07-2075N-201 07-2075R-201 07-3020B-300 07-3020G-300 07-3020N-300 07-3020R-300 07-3021B-300 07-3021G-300 07-3021N-300 07-3021R-300 07-3045B-300 07-3045G-300 07-3045N-300 07-3045R-300 07-3075B-300 07-3075G-300 07-3075N-300 07-3075R-300 07-8020B-1200 07-8020B-2200 07-8020G-1200 07-8020G-2200 07-8020N-1200 07-8020N-2200 07-8020R-1200 07-8020R-2200 07-8021B-1200 07-8021B-2200 07-8021G-1200 07-8021G-2200 07-8021N-1200 07-8021N-2200 07-8021R-1200 07-8021R-2200 07-8045B-1200 07-8045B-2200 07-8045G-1200 07-8045G-2200 07-8045N-1200 07-8045R-1200 07-8045R-2200 07-8075B-1200 07-8075B-2200 07-8075G-1200 07-8075G-2200 07-8075N-1200 07-8075N-2200 07-8075R-1200 07-8075R-2200 07-1270B-101 07-1270G-101 07-1270N-101 07-1270R-101 07-2070B-201 07-2070G-201 07-2070N-201 07-2070R-201 07-3070B-300 07-3070G-300 07-3070N-300 07-3070R-300 07-8070B-1200 07-8070B-2200 07-8070G-1200 07-8070G-2200 07-8070N-1200 07-8070N-2200 07-8070R-1200 07-8070R-2200 07-3070B-350 07-3070G-350 07-3070N-350 07-3070R-350 07-3075B-350 07-3075G-350 07-3075N-350 07-3075R-350 07-2020B-250 07-2020G-250 07-2020N-250 07-2020R-250 07-2021B-250 07-2021G-250 07-2021N-250 07-2045B-250 07-2045G-250 07-2045N-250 07-2045R-250 07-2070B-250 07-2070G-250 07-2070N-250 07-2070R-250 07-2075B-250 07-2075G-250 07-2075N-250 07-2075R-250 07-3020B-350 07-3020G-350 07-3020N-350 07-3020R-350 07-3021B-350 07-3021G-350 07-3021N-350 07-3021R-350 07-3045B-350 07-3045G-350 07-3045N-350 07-3045R-350
97044-954EA 210.84 CAD
97044-954 97044-958 97044-970 97044-974 97044-978 97044-982 97044-994 97044-998 97045-002 97045-006 97045-018 97045-022 97045-032 97045-036 97045-048 97045-052 97045-068 97045-072 97045-084 97045-088 97045-092 97045-096 97045-108 97045-112 97045-116 97045-120 97045-134 97045-138 97045-148 97045-152 97045-164 97045-168 97045-198 97045-202 97045-214 97045-218 97045-222 97045-226 97045-238 97045-242 97045-246 97045-250 97045-264 97045-268 97045-278 97045-282 97045-294 97045-298 97045-384 97045-392 97045-396 97045-400 97045-404 97045-412 97045-416 97045-420 97045-464 97045-472 97045-476 97045-480 97045-484 97045-492 97045-496 97045-500 97045-504 97045-512 97045-516 97045-520 97045-524 97045-532 97045-536 97045-540 97045-584 97045-592 97045-596 97045-600 97045-604 97045-612 97045-616 97045-620 97045-624 97045-632 97045-636 97045-640 97045-644 97045-652 97045-656 97045-660 97045-704 97045-712 97045-716 97045-724 97045-732 97045-740 97045-748 97045-756 97045-760 97045-764 97045-768 97045-776 97045-780 97045-784 97045-828 97045-836 97045-840 97045-844 97045-848 97045-856 97045-860 97045-864 89231-778 89231-782 89231-792 89231-796 89231-800 89231-804 89231-816 89231-820 89231-824 89231-828 89231-840 89231-844 89231-848 89231-856 89231-860 89231-864 89231-868 89231-876 89231-880 89231-884 89231-928 89231-936 89231-940 89231-944 89231-948 89231-956 89231-960 89231-964 30618-400 30618-404 30618-416 30618-420 30618-424 30618-428 30618-440 30618-444 30618-206 30618-210 30618-222 30618-226 30618-230 30618-234 30618-246 30618-250 30618-254 30618-258 30618-270 30618-274 30618-278 30618-282 30618-294 30618-298 30618-302 30618-306 30618-318 30618-322 30618-328 30618-332 30618-344 30618-348 30618-352 30618-356 30618-368 30618-372 30618-376 30618-380 30618-392 30618-396
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Inserts and Snap Caps, Unassembled, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with glass inserts and color-coded caps.
  • Variety of volumes available
  • Standard chromatography closures
  • Snap caps
  • Silicone mats are resealable
A comprehensive line of 96-Well microplates for chromatography, the Plate Sampling System™ uses closures familiar to those who work with standard chromatography vials
Polypropylene μLPlates® have a variety of closures that will stabilize the sample during the analytical sample run and, if necessary,More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable