WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Covers, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
07-1010N 07-1063N 07-1065N 07-1066N 07-1110N 07-1163N 07-1165N 07-1166N 07-1210N 07-1263N 07-1265N 07-1266N 07-2010N 07-2063N 07-2065N 07-2066N 07-3010N 07-3063N 07-3065N 07-3066N 07-5015N 07-5017N 07-5060N 07-5061N 07-5067N 07-6015N 07-6017N 07-6060N 07-6061N 07-6067N 07-7015N 07-7017N 07-7060N 07-7061N 07-7067N 07-8015N 07-8017N 07-8060N 07-8061N 07-8067N 07-3963N 07-3965N 07-3910N
97044-918EA 25.6 CAD
97044-918 97044-920 97044-924 97044-926 97044-928 97044-932 97044-934 97044-938 97044-940 97044-942 97044-948 97044-950 97045-026 97045-028 97045-030 97045-062 97045-064 97045-142 97045-144 97045-146 97045-176 97045-180 97045-182 97045-184 97045-192 97045-194 97045-272 97045-274 97045-276 97045-324 97045-326 97045-328 97045-329 97045-330 97045-334 97045-336 97045-338 97045-339 97045-340 97045-344 97045-346 97045-348 97045-349 97045-350 97045-356 97045-358 97045-360 97045-361 97045-362 97045-380 97045-382 97045-744 97045-745 97045-746 97045-872 97045-874 97045-876 97045-877 97045-878 10806-034 10806-054 10806-070 10806-072
WHEATON® MicroLiter µLPlate® Component Kits, PP Plates with Covers, DWK Life Sciences
Microplates Deep Well Microplates
MicroLiter µLPlate® components include a proprietary product line of 96-well round and square well microplates with covers.
  • Variety of volumes available
  • EVA covers offer superior seal
  • Silicone mats are resealable
The 96-well round and square well microplates are available in a variety of volumes. Constructed of natural polypropylene, all microplates are made to ANSI specifications for 96-well microplates. Covers made of Ethylene Vinyl Acetate (EVA) are piercible by standard needle configurations found on most HPLC, LC/MS, CombiChem, and HTSMore Product Information