Economical Individual Weights, Troemner
Supplier: Troemner
30391015 30391012 30391009 30391006 30391003 30391000 30391014 30391011 30391008 30391005 30391002 30390999 30391013 30391010 30391007 30391004 30391001 30391016 30391017 30391018
97044-294EA 45.68 CAD
97044-294 97044-296 97044-298 97044-300 97044-302 97044-304 97044-306 97044-308 97044-310 97044-312 97044-314 97044-316 97044-318 97044-320 97044-322 97044-324 97044-326 10754-344 10754-346 10754-348
Economical Individual Weights, Troemner
Weights Calibration Weights
Individual weights are available from 2 kg to 1 mg.
Weights 1 g and above are carefully manufactured of stainless steel and are cylindrical in shape for easy gripping. Weights 1 g to 100 g are made from one-piece construction and are adjusted during manufacture by removing material from the bottom of the weight.
Weights 200 g through 2 kg have an adjusting cavity on the top that is sealed with a plug. Economical stainless steel weights 1 g and above are perfectly flat on top to stack easily on center.
Weights 500 mg and lower are made of sheet metal. They feature one-pieceMore Product Information