Electrolyte Solution, L5014, SI Analytics
Supplier: YSI
285138324
97042-456EA 337.05 CAD
97042-456
Electrolyte Solution, L5014, SI Analytics
Electrolytes Electrode Filling Solutions
Acetic acid saturated with lithium chloride.