Metal Combination Electrodes, SI Analytics
Supplier: SI Analytics
285102343 285102413 285102208
97041-820EA 2037.82 CAD
97041-820 97041-856 97041-858 97042-002 97042-004
Metal Combination Electrodes, SI Analytics
Electrodes Other Electrodes
Metal combination electrodes with plug head and connection cable with silver/silver chloride reference system.
Order Now
SPECIFICATIONS
Environmentally Preferable