EMF Meter, Sper Scientific
Supplier: Sper Scientific
840045
97035-938EA 263.97 CAD
97035-938
EMF Meter, Sper Scientific
Electromagnetic Field Meters
Measures electromagnetic field emissions from electrical power lines and transmission equipment, computer, HVAC, audio/video, and other electrical appliances and equipment.
While the evidence of the carcinogenic effect of such electromagnetic emissions is still inconclusive, prudent avoidance is recommended to avoid excess exposure over long periods of time. Reads milli-Gauss and micro-Tesla on a large 1¼" high display. The EMF Meter's display indicates when the object being measured is out of range as well as when the battery is low. Features both standard and maximum hold. Powered by aMore Product Information