Psychrometer/Anemometer, Sper Scientific
Supplier: Sper Scientific
840034
97035-936EA 533.71 CAD
97035-936
Psychrometer/Anemometer, Sper Scientific
Hygrometers Psychrometers
This unique instrument combines the features of an anemometer and a psychrometer: Air Speed, Air Volume, BTU (British Thermal Units), Air Temperature, Dew Point, Wet Bulb, Humidity.
Also calculates air volume and BTU capacity. Excellent for HVAC, IAQ and environmental work. Auto ranging with air velocity in m/s or ft/s, air volume in CFM or m3/sec and temperature in °C or °F. Features min/max/ave, hold, auto power off, LCD backlight and a computer interface. The probe detaches for easy replacement and repair.
Ordering information: 5-year warranty. Comes in a hard carrying case completeMore Product Information