YMC-Pack C4 (Butyl) HPLC Columns, YMC America
Supplier: YMC America,Inc
BU12S05-0502WT BU12S03-1002WT BU12S03-1003WT BU12S03-1046WT BU12S03-1502WT BU12S03-1503WT BU12S03-1546WT BU12S05-0310WTG BU12S05-0520WTG BU12S05-0530WTG BU12S05-1006WT BU12S05-1020WT BU12S05-1030WT BU12S05-1046WT BU12S05-1501WT BU12S05-1502WT BU12S05-1506WT BU12S05-1510WT BU12S05-1520WT BU12S05-1530WT BU12S05-1546WT BU12S05-2501WT BU12S05-2502WT BU12S05-2506WT BU12S05-2510WT BU12S05-2520WT BU12S05-2530WT BU12S05-2546WT BU12S05-3006WT BU12S05-3010WT BU12S05-3046WT BU12S05-L530WT BU12S05-L546WT BU20S05-0520WTG BU20S05-0530WTG BU20S05-1520WT BU20S05-1546WT BU20S05-2520WT BU20S05-2530WT BU20S05-2546WT BU30S05-0310WTG BU30S05-0520WTG BU30S05-0530WTG BU30S05-1006WT BU30S05-1046WT BU30S05-1501WT BU30S05-1502WT BU30S05-1506WT BU30S05-1510WT BU30S05-1520WT BU30S05-1546WT BU30S05-2501WT BU30S05-2502WT BU30S05-2506WT BU30S05-2510WT BU30S05-2520WT BU30S05-2530WT BU30S05-2546WT BU30S05-3006WT BU30S05-3010WT BU30S05-3046WT BU30S05-L546WT BU12S05-0503WT BU12S05-0504WT BU12S05-0546WT BU20S05-0502WT BU20S05-0503WT BU20S05-0504WT BU30S05-0502WT BU30S05-0503WT BU30S05-0504WT BU30S05-0546WT
89198-940EA 1152.56 CAD
89198-940 97029-770 97029-772 97029-776 97029-778 97029-780 97029-782 97029-812 97029-820 97029-822 97029-828 97029-830 97029-832 97029-836 97029-838 97029-840 97029-842 97029-844 97029-846 97029-848 97029-850 97029-852 97029-854 97029-856 97029-858 97029-860 97029-862 97029-864 97029-866 97029-870 97029-872 97029-874 97029-876 97029-884 97029-886 97030-000 97030-002 97030-006 97030-008 97030-010 97030-012 97030-014 97030-092 97030-100 97030-102 97030-108 97030-112 97030-114 97030-116 97030-118 97030-120 97030-122 97030-124 97030-126 97030-128 97030-130 97030-132 97030-134 97030-136 97030-138 97030-140 97030-144 97030-146 97030-148 97030-150 97030-158 89216-586 89216-588 89216-590 89216-606 89216-608 89216-610 89216-616 89216-618 89216-620 89216-622
YMC-Pack C4 (Butyl) HPLC Columns, YMC America
Chromatography Columns
YMC-Pack C4 (butyl) columns are less hydrophobic than C8 or C18 packings, and generally utilize more aqueous eluents than either of these reversed-phase column types.
  • Fully endcapped
  • Ideally suited for biological separations
  • High-coverage monomeric bonding on perfectly spherical silica supports
  • Available in 120, 200 and 300 Å pore size packings
When compared to C8 and C18 columns (using the same eluents), C4 stationary phases show significantly shorter retention for non-polar compounds. Retention of polar compounds, however, is not significantlyMore Product Information