HPLC Columns, YMC-Pack ODS-A
Supplier: YMC America,Inc
AA12S05-0546WT AA12S03-0104WFG AA12S03-0105WFG AA12S03-0502WT AA12S03-0503WT AA12S03-0504WT AA12S03-0546WT AA12S03-1002WT AA12S03-1003WT AA12S03-1006WT AA12S03-1046WT AA12S03-1502WT AA12S03-1503WT AA12S03-1504WT AA12S03-1506WT AA12S03-1546WT AA12S03-2546WT AA12S03-H502WT AA12S03-L502WT AA12S03-L546WT AA12S05-0104WFG AA12S05-0105WFG AA12S05-0310WTG AA12S05-0520WTG AA12S05-0530WTG AA12S05-05Q1WT AA12S05-1006WT AA12S05-1020WT AA12S05-1030WT AA12S05-1046WT AA12S05-1501WT AA12S05-1502WT AA12S05-1503WT AA12S05-1506WT AA12S05-1510WT AA12S05-1520WT AA12S05-1530WT AA12S05-1546WT AA12S05-15Q1WT AA12S05-2501WT AA12S05-2502WT AA12S05-2503WT AA12S05-2506WT AA12S05-2510WT AA12S05-2520WT AA12S05-2530WT AA12S05-2546WT AA12S05-3006WT AA12S05-3010WT AA12S05-3046WT AA12S05-L502WT AA12S05-L530WT AA12S05-L546WT AA12S11-0104WFG AA12S11-0105WFG AA12S11-0310WTG AA12S11-0520WTG AA12S11-0530WTG AA12S11-1006WT AA12S11-1046WT AA12S11-1506WT AA12S11-1510WT AA12S11-1520WT AA12S11-1546WT AA12S11-2506WT AA12S11-2510WT AA12S11-2520WT AA12S11-2530WT AA12S11-2546WT AA12S11-3006WT AA12S11-3010WT AA12S11-5020WT AA12S11-5030WT AA30S05-0104WFG AA30S05-0105WFG AA30S05-0310WTG AA30S05-0520WTG AA30S05-0530WTG AA30S05-05Q1WT AA30S05-1006WT AA30S05-1046WT AA30S05-1501WT AA30S05-1502WT AA30S05-1506WT AA30S05-1510WT AA30S05-1520WT AA30S05-1546WT AA30S05-15Q1WT AA30S05-2501WT AA30S05-2502WT AA30S05-2506WT AA30S05-2510WT AA30S05-2520WT AA30S05-2530WT AA30S05-2546WT AA30S05-3006WT AA30S05-3010WT AA30S05-3046WT AA30S05-L502WT AA30S05-L546WT AA12S03-01Q1GC AA12S03-0204GC AA12S05-01Q1GC AA12S05-0204GC AA12S05-02Q1GC AA12S05-0502WT AA12S05-0503WT AA12S05-0504WT AA30S05-0204GC AA30S05-02Q1GC AA30S05-0502WT AA30S05-0503WT AA30S05-0504WT AA12S03-02Q1GC
89198-928EA 1443.58 CAD
89198-928 97028-004 97028-006 97028-016 97028-020 97028-024 97028-028 97028-030 97028-032 97028-034 97028-038 97028-040 97028-042 97028-044 97028-046 97028-048 97028-050 97028-060 97028-062 97028-064 97028-072 97028-074 97028-084 97028-092 97028-096 97028-102 97028-104 97028-106 97028-110 97028-114 97028-116 97028-118 97028-120 97028-122 97028-124 97028-126 97028-128 97028-130 97028-132 97028-134 97028-136 97028-138 97028-140 97028-142 97028-144 97028-146 97028-148 97028-150 97028-154 97028-156 97028-158 97028-160 97028-170 97028-172 97028-174 97028-182 97028-184 97028-186 97028-190 97028-192 97028-196 97028-198 97028-200 97028-202 97028-204 97028-206 97028-208 97028-210 97028-212 97028-214 97028-216 97028-220 97028-222 97028-226 97028-228 97028-422 97028-424 97028-434 97028-442 97028-444 97028-448 97028-450 97028-452 97028-454 97028-456 97028-458 97028-460 97028-462 97028-464 97028-466 97028-468 97028-470 97028-472 97028-474 97028-476 97028-478 97028-480 97028-482 97028-486 97028-488 97028-490 97028-492 97028-498 97028-500 97028-502 97028-504 97028-506 97028-508 97028-510 97028-514 97028-516 97028-518 97028-520 97028-522 97028-524 97028-526 97028-528 97028-530 97028-532 97028-534 97028-538 97028-540 97028-542 97028-544 97028-546 89216-370 89216-372 89216-378 89216-380 89216-382 89216-384 89216-386 89216-388 89216-420 89216-422 89216-424 89216-426 89216-428 89216-374
HPLC Columns, YMC-Pack ODS-A
Chromatography Columns
ODS-A is a high coverage, fully endcapped C18 phase which exhibits exceptional batch-to-batch reproducibility.
  • Highly versatile ODS phase
  • Selectivity for polar to moderately nonpolar pharmaceuticals, organic chemicals, biologicals and natural products
ODS-A is the first choice of many chromatographers for high performance reversed phase separations. It has selectivity for the separation of a wide range of organic compounds found in pharmaceutical, chemical and biotechnology industries. ODS-A is a high coverage, fully endcapped C18 phase which exhibitsMore Product Information