XPert™ Nano Ventilated Enclosures, Labconco®
Supplier: Labconco
3887220 3887221 3887260 3887261 3887262 3887320 3887321 3887360 3887361 3887362 3887420 3887421 3887460 3887461 3887462 3887520 3887521 3887560 3887561 3887562 3887620 3887621 3887660 3887661 3887662 3888520 3888521 3888560 3888561 3888562 3888620 3888621 3888660 3888661 3888662
97025-424EA 15223.3 CAD
97025-424 97025-426 97025-428 97025-430 97025-432 97025-434 97025-436 97025-438 97025-440 97025-442 97025-444 97025-446 97025-448 97025-450 97025-452 97025-454 97025-456 97025-458 97025-460 97025-462 97025-464 97025-466 97025-468 97025-470 97025-472 97025-474 97025-476 97025-478 97025-480 97025-482 97025-484 97025-486 97025-488 97025-490 97025-492
XPert™ Nano Ventilated Enclosures, Labconco®
Fume Hoods and Enclosures Extraction Cabinets
During procedures such as nanoparticle manipulation and dry powder chemical handling, enclosures keep hazardous powders and particulates contained
Features deep interiors, Guardian™ airflow monitor, true bag-in/bag-out filter disposal system, built-in blower, ergonomic air foil with aerodynamic Clean-Sweep™ openings that enhance airflow, upper dilution air supply, upper containment sash foil, zone-perforated rear baffle, and pressure gauge for monitoring filter loading. Type 304 stainless steel interior sides, removable baffle, removable air foil, and integral work surface are easy toMore Product Information