iLog® Temperature Loggers, Cryopak Verification Technologies
Supplier: Cryopak Verification Technologies
EI-1E-DC-32-L EI-1E-D-32-L EI-2E-D-32-L EI-2E-C-32-L EI-IN-D-32-L EI-2E-N-32-L EI-2E-Z-32-L EI-2E-DC-32-L EI-2E-DN-32-L EI-2E-DZ-32-L EI-2E-CN-32-L EI-2E-CZ-32-L EI-2E-NZ-32-L
97017-198EA 713.5 CAD
97017-198 97017-200 97017-204 97017-216 97017-238 10033-852 10033-854 10033-856 10033-858 10033-860 10033-862 10033-864 10033-866
iLog® Temperature Loggers, Cryopak Verification Technologies
Data Loggers
The user programmable iLog data logger provides technological abilities for monitoring temperature sensitive products while supporting compliance requirements. The case is made from Polycarbonate.
  • Large LCD display with multi-functional viewing
  • Visual and audible alarms
  • Bookmark capabilities
  • Wide temperature range (high heat, ambient, refrigerated, frozen and dry ice)
  • High alarm; low alarm configuration
  • Two year warranty
Detachable, customizable external sensor lengths and probe styles.
Certifications: ISO9001:2008, CE, RoHS, EN 12830.