Orion™ Green pH Combination Electrodes, Thermo Scientific
Supplier: Thermo Fisher Scientific
GS9106BNWP GD9156BNWP GD9106BNWP
97014-782EA 273.22 CAD
97014-782 97014-788 97014-784
Orion™ Green pH Combination Electrodes, Thermo Scientific
Electrodes pH Electrodes
The Green electrodes are the first laboratory glass-bulb pH electrodes to meet all the requirements of RoHS.
  • Choice of single-junction models for clean-water measurements, or double-junction models for dirty-water testing
  • Choice of refillable or low-maintenance gel-filled models
  • No disposal issues or disposal cost
  • pH range is 0 to 14
  • Durable epoxy body
These environmentally friendly pH combination electrodes with epoxy body and waterproof BNC connector contain no lead, mercury or other hazardous substance.