Digital Dry Block Heater, QBD/QBH, Grant Instruments
Supplier: GRANT USA INC.
QBD2L QBH2L
97011-886EA 2706.22 CAD
97011-886 97011-890
Digital Dry Block Heater, QBD/QBH, Grant Instruments
Block Heaters Digital Block Heaters
Compact digital block heaters with choice of models for 1 or 2 or 4 interchangeable blocks.
  • Stability and uniformity ±0.1ºC
  • Digitsl temperature control for optimum precision
  • Heating range: ambient +5ºC to 130ºC, with rapid heat-up time
  • Range of convenient features including alarms, single and dual point calibration, programmed start/stop, 'offset' for known sample temperature variation and choice of external or internal probes
  • External probe available for accurate temperature control in a tube
Units feature an LED temperature display, easy to useMore Product Information