MaxQ™ 8000 Stackable Orbital Shakers, Thermo Scientific
Supplier: Thermo Fisher Scientific
SHKE8000 SHKE8000-7 SHKE8000-7PROMO SHKE8000PROMO
97011-462EA 37130 CAD
97011-462 97011-466 76266-330 76266-332
MaxQ™ 8000 Stackable Orbital Shakers, Thermo Scientific
Shakers Orbital Shakers
Stackable orbital shakers provide an ideal solution for both sensitive cultures and traditional 'hard shake' applications in incubated and refrigerated configurations.
A heavy-duty, eccentric drive mechanism allows extended speed ranges with minimal vibration, even when units are stacked. The slide-out platform features a large mounting surface area and is secured to ensure smooth and stable motion. Platform motion is continually monitored independently of motor operation. The cool running, brushless, ¹/₃ HP, DC motor provides maintenance-free operation and long life. The horizontal HEPAMore Product Information