VWR® CryoGuard Cryogenic Gloves
Supplier: VWR International
97008-196 97008-198 97008-200 97008-202 97008-204 97008-206 97008-208 97008-210 97008-212 97008-216 97008-218 97008-220 97008-222 97008-224 97008-226 97008-228 97008-230 97008-232 97008-234 97008-236 97008-242 97008-244 97008-248 97008-250 97008-252 97008-254 97008-256 97008-258 97008-214 97008-238 97008-240 97008-246
97008-196EA 196.67 CAD
97008-196 97008-198 97008-200 97008-202 97008-204 97008-206 97008-208 97008-210 97008-212 97008-216 97008-218 97008-220 97008-222 97008-224 97008-226 97008-228 97008-230 97008-232 97008-234 97008-236 97008-242 97008-244 97008-248 97008-250 97008-252 97008-254 97008-256 97008-258 97008-214 97008-238 97008-240 97008-246
VWR® CryoGuard Cryogenic Gloves
Gloves
Multi-layer, insulated gloves are lightweight, flexible, and durable
  • Multi-layer and insulated
  • Lightweight, flexible, and durable
  • Moisture-wicking liner
  • Waterproof
  • Various lengths available
Multi-layer, insulated gloves are designed to protect hands and arms from the hazards of working with cryogenic materials. They can be worn for extended periods of time, as they are extremely warm, even in temperatures as low as –196°C. Water-resistant and waterproof gloves feature two layers of insulation material bonded together at the edges to trap insulatingMore Product Information