Class 1 and UltraClass Cal-Paks with NVLAP Calibration Certificate, Troemner
Supplier: Troemner
30391799 30390007 30391795 30391794 30391793 30391792 30391790 30391789 30391788 30391787 30391786 30391785 30391784 30391783 30390006 30391782 30391798 30391791
97007-962EA 1628.1 CAD
97007-962 97007-970 97007-974 97007-978 97007-982 97007-986 97007-994 97007-998 97008-002 97008-006 97008-010 97008-014 97008-018 97008-022 97008-026 97008-030 97007-966 97007-990
Class 1 and UltraClass Cal-Paks with NVLAP Calibration Certificate, Troemner
Weights Calibration Weights
Troemner Cal-Paks consist of high quality precision mass standards with the lowest measurement uncertainties, packaged into a versatile weight case
Cal-Paks enable users to meet all regulatory and audit needs, including ISO, FDA, GMP, GLP, DOD, ANSI/NCSL Z540-1, 10CFR50, 21CFR, and USP 41. Each Cal-Pak contains three ASTM Alloy 8 Class 1 or UltraClass weights. The trio of weights include the manufacturer's recommended calibration weight, 10% of the calibration weight, and the recommended minimum weight determined by the balance's readability and published standard deviation under normalMore Product Information