Paramount® and Paramount 360® Ductless Enclosures, Labconco®
Supplier: Labconco
6963200 6963220 6963300 6963301 6963302 6963320 6963321 6963322 6963400 6963401 6963402 6963403 6963420 6963421 6963422 6963423 6963501 6963503 6963521 6963523 6963601 6963603 6963621 6963623 6938200 6938201 6938202 6938203 6938100
97007-602EA 11834.2 CAD
97007-602 97007-604 97007-606 97007-608 97007-610 97007-612 97007-614 97007-616 97007-618 97007-620 97007-622 97007-624 97007-626 97007-628 97007-630 97007-632 97007-634 97007-636 97007-638 97007-640 97007-642 97007-644 97007-646 97007-648 97007-650 97007-652 97007-654 97007-656 97007-662
Paramount® and Paramount 360® Ductless Enclosures, Labconco®
Fume Hoods and Enclosures Ductless Fume Hoods
Self-contained ductless enclosures allow safe handling of organic, formaldehyde, acid gas, sulfur compounds, or ammonia chemical fumes and vapors.
  • Upper Containment Sash Foil
  • Upper Dilution Air Supply
  • Slotted Rear Baffle
  • Enclosures Feature 360° Visibility
  • Feet Can Be Replaced with Casters for Mobility
Enclosures feature an upper containment sash foil, upper dilution air supply, and a slotted rear baffle that promotes horizontal laminar airflow by drawing vapors and fumes to the back of the enclosure without causing turbulence that can impedeMore Product Information