Base Stands and Accessories for Purifier® Logic® Series Class II Safety Cabinets, Labconco®
Supplier: Labconco
3747500 3730310 3730410 3780200 3780201 3780202 3780101 3780102 3780103 3780104 3780105 3784000 3780310 3780311 3780312 3780313 3830510 3780203 3780106 3780107 3811105 3811106 3811107 3811108 3852000 3859000 3850100 3850200 3858800 3859100 7071900 3859600 3859602 3859603 3859601 3858600 3858603 3858602 3858601 3858500 3858501 3858502 3859400P 3857000
43100-012EA 2346.08 CAD
43100-012 46610-764 82003-576 82003-578 82003-588 82003-590 82003-592 82003-606 82003-608 82003-610 82003-612 82003-614 82003-616 82003-630 82013-190 82013-194 82013-198 82013-202 89003-088 89003-090 89003-092 89003-094 89003-096 97006-422 97006-424 97006-426 97006-428 97006-950 97006-952 97006-954 97006-956 97006-958 97006-960 97006-966 97006-984 97006-986 97007-000 97007-002 97007-004 97007-006 97007-016 97007-018 97007-028 97007-030 97007-032 97007-034 97007-036 97007-038 97007-040 97007-042 97007-594 97007-596 97010-022 82003-580 83009-284
Base Stands and Accessories for Purifier® Logic® Series Class II Safety Cabinets, Labconco®
Fume Hoods and Enclosures PCR Workstation Enclosures
Logic Cart fits under any telescoping base stand. Features three small and one large compartments, one drawer, front panel cut out for dispensing wipers or gloves, and HDPE worksurface. W×D×H: 635×483×686 mm (25×19×27").
Service Fixture Kit includes serrated hose tip valve with ball valve, quarter turn control handle, hardware and instructions for plumbing to services. Mounts on left or right side interior. Up to two may be installed per side.
Base Stands — Constructed of durable 45 mm (1¾") tubular, glacier white epoxy-coated steel, these ADA-compliant base stands support up to 453.6More Product Information