Intrinsic Dissolution Accessories, Quality Lab Accessories
Supplier: QLA
IDADIE-01 IDAGST-01 IDAPCH-01 IDAPLT-01 IDASET-01 IDASNH-01 IDASNH-ST IDASET-EW IDASET-ST IDASNH-EW
97002-376EA 592.5 CAD
97002-376 97002-378 97002-380 97002-382 97002-386 97002-388 75848-942 75849-232 75849-234 75849-236 75849-238 75849-240
Intrinsic Dissolution Accessories, Quality Lab Accessories
Dissolution Apparatus
QLA manufactures Intrinsic Dissolution accessories for use on many dissolution baths.
  • The QLA intrinsic dissolution apparatus is based on the conventional "wood apparatus design"
This industry standard configuration has a long and successful history. A competitive configuration known as the "Rotating Disk System" requires a flat bottom dissolution vessel. The QLA design uses standard dissolution vessels and provides better centering and easier dosage placement.
Order Now
SPECIFICATIONS
Environmentally Preferable