Sure-Grip® EX Safety Cabinets for Flammable Materials, Justrite®
Supplier: Justrite
893020 891720 890400 891300 892300 891500 893400 8930053 8930253 894500 896000 899000 891220 896020 892320 891700 890420 891200 891520 891320 8945253 8945053 894580 896080 899080 893300 894520 899020 890500
97001-210EA 2694.43 CAD
97001-210 97001-238 97001-248 97001-342 97001-352 97001-362 97001-364 97001-366 97001-372 97001-384 97001-400 97006-112 97006-114 97006-116 97006-118 97006-120 97006-122 97006-126 97006-128 97006-130 97006-140 97006-142 97033-754 97049-636 97049-640 97001-368 97001-390 97001-410 76018-056
Sure-Grip® EX Safety Cabinets for Flammable Materials, Justrite®
Furniture Storage Cabinets
Countertop and Compac cabinets provide safe storage when space is limited
  • Fire-resistant, welded 18-gauge steel
  • Double-wall design features a 3.8 cm (1/2”) insulating air space
  • Built-in grounding connector prevents static buildup
  • Adjustable shelves
Fire-resistant, welded 18-gauge steel cabinets store flammable liquids and waste safely and securely, protecting personnel and facilities. The double-wall design features a 3.8cm (11/2") insulating air space. A 3-point stainless steel bullet latching system further reduces the risk of fire. ExclusiveMore Product Information