UltraMOP™ Mop Head for Cleanroom Environments, Foamtec International
Supplier: Foamtec International
FS600ST-12 FS505-B1
89135-142CS 639.57 CAD
89135-142 94024-538
UltraMOP™ Mop Head for Cleanroom Environments, Foamtec International
Mops
Perfex®-style cleanroom foam mop head is designed to fit the TruCLEAN® mopping system
Mop head is constructed of a snag-resistant, polyester fabric lamination that extends over the leading and trailing edges of the underlying foam sponge, eliminating the need for separate polyester socks. This unique Bump-PROOF® design improves cleanliness and durability. Compatible with Gamma, ETO, and autoclave sterilization
Certifications: Lot certified, individually packed, and lot traceable back to raw materials.
Ordering information: Handle and frame not included. Fits Perfex-style mop frameMore Product Information