NMC Lockout Tags
Supplier: ACCUFORM MANUFACTURING, INC
LOTAG13 LOTAG1 LOTAG10 LOTAG11 LOTAG12 LOTAG15 LOTAG16 LOTAG17 LOTAG20 LOTAG21 LOTAG22 LOTAG24 LOTAG27 LOTAG28 LOTAG30 LOTAG31 LOTAG32 LOTAG33 LOTAG34 LOTAG35 LOTAG8 LOTAG9 LOTAG18 LOTAG14
36434-626PK 105.73 CAD
36434-626 56607-832 94016-430 94016-432 94016-434 94016-438 94016-440 94016-442 94016-448 94016-450 94016-452 94016-454 94016-456 94016-458 94016-460 94016-464 94016-466 94016-468 94016-470 94016-472 94016-474 94016-476 94016-478 94016-480 94016-444 89186-006
NMC Lockout Tags
Security Locks
Prevent accidental or unexpected start up or release of stored energy when working on equipment and machinery
  • Prevent accidental or unexpected start-up or release of stored energy
  • Unrippable vinyl, features .95cm (⅜") reinforced brass grommet
Made from Unrippable vinyl, these tags feature a .95cm (⅜") reinforced brass grommet.
Order Now
SPECIFICATIONS
Environmentally Preferable