NMC Large Plug Lockout
Supplier: ACCUFORM MANUFACTURING, INC
HLD
94015-936EA 141.88 CAD
94015-936
NMC Large Plug Lockout
Security Locks
Accommodates plugs up to 7.62cm (3") diameter and 13.97c (5½") long with a maximum cord diameter of 3.18cm (1¼").